Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
APMAP
Entrez Gene ID
416715
Gene Name
adipocyte plasma membrane associated protein
Gene Aliases
BSCv, C20orf3, C3H20ORF3
Chromosome Location
Chr.3: 15927402 - 15940460 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006177.1 NP_001006177.1 6 - 7 721 91
XM_025148593.1 XP_025004361.1 6 - 7 774 91
GenBank mRNA AJ720833.1 - 6 - 7 721 91
CR389678.1 - 6 - 7 720 91

View Full Product Details