Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMPO
Entrez Gene ID
417923
Gene Name
thymopoietin
Gene Aliases
-
Chromosome Location
Chr.1: 46668937 - 46691164 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006235.1 NP_001006235.1 6 - 7 935 121
XM_015284619.2 XP_015140105.1 6 - 7 979 121
XM_015284625.2 XP_015140111.1 6 - 7 978 121
XM_015284631.2 XP_015140117.1 5 - 6 861 121
XM_015284646.2 XP_015140132.1 5 - 6 856 121
GenBank mRNA AJ719348.1 - 6 - 7 935 121

View Full Product Details