Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ING4
Entrez Gene ID
418281
Gene Name
inhibitor of growth family member 4
Gene Aliases
-
Chromosome Location
Chr.1: 77735015 - 77749824 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006251.1 NP_001006251.1 5 - 6 528 85
XM_015293359.2 XP_015148845.1 6 - 7 701 85
GenBank mRNA AJ719950.1 - 5 - 6 528 85

View Full Product Details