Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPRYD7
Entrez Gene ID
418871
Gene Name
SPRY domain containing 7
Gene Aliases
C13orf1, C1H13orf1, CLLD6
Chromosome Location
Chr.1: 170674866 - 170683661 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006273.1 NP_001006273.1 2 - 3 304 77
GenBank mRNA AJ721027.1 - 2 - 3 304 77
CR385289.1 - 2 - 3 273 77

View Full Product Details