Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BDH1A
Entrez Gene ID
424891
Gene Name
3-hydroxybutyrate dehydrogenase 1A
Gene Aliases
BDH, BDH1
Chromosome Location
Chr.9: 12794246 - 12807839 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006547.2 NP_001006547.2 3 - 4 313 162
XM_015291389.2 XP_015146875.1 4 - 5 365 162
XM_025153361.1 XP_025009129.1 4 - 5 468 162
GenBank mRNA AJ720289.1 - 4 - 5 320 150

View Full Product Details