Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BMP15
Entrez Gene ID
428697
Gene Name
bone morphogenetic protein 15
Gene Aliases
GDF-9B
Chromosome Location
Chr.4: 1860090 - 1862633 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006589.2 NP_001006589.2 1 - 2 652 140
GenBank mRNA AY725199.1 - 1 - 2 652 140

View Full Product Details