Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBXW2
Entrez Gene ID
417088
Gene Name
F-box and WD repeat domain containing 2
Gene Aliases
-
Chromosome Location
Chr.17: 10673253 - 10682820 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007842.1 NP_001007843.1 5 - 6 1262 145
XM_015279379.2 XP_015134865.1 5 - 6 2054 145
XM_015279381.2 XP_015134867.1 5 - 6 2015 145
XM_015279382.2 XP_015134868.1 5 - 6 1347 145
XM_015279383.2 XP_015134869.1 5 - 6 1365 145
XM_015279385.2 XP_015134871.1 5 - 6 1255 145
XM_025156037.1 XP_025011805.1 5 - 6 1224 145
GenBank mRNA AJ720209.1 - 5 - 6 1262 145

View Full Product Details