Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM230
Entrez Gene ID
419518
Gene Name
transmembrane protein 230
Gene Aliases
C20orf30, C22H20ORF30
Chromosome Location
Chr.22: 484294 - 499838 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007877.1 NP_001007878.1 4 - 5 455 137
GenBank mRNA AJ719760.1 - 4 - 5 455 137

View Full Product Details