Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DCLRE1A
Entrez Gene ID
423902
Gene Name
DNA cross-link repair 1A
Gene Aliases
SNM1A
Chromosome Location
Chr.6: 28504696 - 28519260 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008683.1 NP_001008683.1 3 - 4 2047 126
XM_015288657.1 XP_015144143.1 3 - 4 2582 126
GenBank mRNA AY376896.1 - 3 - 4 2047 126

View Full Product Details