Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MEOX1
Entrez Gene ID
395533
Gene Name
mesenchyme homeobox 1
Gene Aliases
GGMOXR1
Chromosome Location
Chr.27: 5939150 - 5944728 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204765.1 NP_990096.1 2 - 3 815 82
GenBank mRNA AF043432.2 - 2 - 3 815 82

View Full Product Details