Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTH
Entrez Gene ID
396436
Gene Name
parathyroid hormone
Gene Aliases
-
Chromosome Location
Chr.5: 7786836 - 7790904 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205452.2 NP_990783.1 2 - 3 201 118
GenBank mRNA EU308591.1 - 1 - 2 85 118
M31604.1 - 2 - 3 203 118
M36522.1 - 2 - 3 212 118

View Full Product Details