Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLIN1
Entrez Gene ID
415487
Gene Name
perilipin 1
Gene Aliases
PLIN
Chromosome Location
Chr.10: 13263491 - 13268056 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127439.1 NP_001120911.1 4 - 5 599 82
XM_015292004.2 XP_015147490.1 5 - 6 687 82
XM_015292005.2 XP_015147491.1 4 - 5 492 82
XM_025153844.1 XP_025009612.1 6 - 7 706 82
GenBank mRNA EU620701.1 - 4 - 5 599 82
GU327532.1 - 5 - 6 672 82

View Full Product Details