Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPIN1W
Entrez Gene ID
374014
Gene Name
spindlin 1-W
Gene Aliases
SPIN, SPIN1, SPINW
Chromosome Location
Chr.W: 5939316 - 6052602 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204191.1 NP_989522.1 5 - 5 2244 109
XM_015300449.2 XP_015155935.1 6 - 6 3293 109
XM_025144527.1 XP_025000295.1 7 - 7 3393 109
GenBank mRNA AB047852.1 - 5 - 5 2244 109

View Full Product Details