Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GJB1
Entrez Gene ID
378797
Gene Name
gap junction protein beta 1
Gene Aliases
CX32
Chromosome Location
Chr.4: 2316357 - 2318810 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204371.2 NP_989702.1 2 - 2 956 62
XM_025149469.1 XP_025005237.1 2 - 2 1029 62
GenBank mRNA AY363307.1 - - 1259 62
BX936196.1 - 2 - 2 956 62

View Full Product Details