Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ADMP
Entrez Gene ID
395622
Gene Name
anti-dorsalizing morphogenetic protein
Gene Aliases
-
Chromosome Location
Chr.28: 993852 - 996088 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204822.2 NP_990153.1 5 - 5 1256 66
GenBank mRNA AF082178.1 - 5 - 5 1257 66

View Full Product Details