Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
HOXC8
Entrez Gene ID
395711
Gene Name
homeobox C8
Gene Aliases
HOXC-8
Chromosome Location
Chr.33: 7612286 - 7615704 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204893.2 NP_990224.1 1 - 1 742 95
GenBank mRNA X94179.1 - - 748 95

View Full Product Details