Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CPP1
Entrez Gene ID
547664
Gene Name
cysteine-rich polycomb-like protein
Gene Aliases
-
Chromosome Location
Chr.17: 34556087 - 34562085 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001249183.1 NP_001236112.1 2 - 3 328 73
GenBank mRNA AJ010165.1 - 2 - 3 328 73

View Full Product Details