Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WIP1
Entrez Gene ID
547675
Gene Name
WIP1 protein
Gene Aliases
-
Chromosome Location
Chr.14: 43919560 - 43922130 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AJ311808.1 - 1 - 2 336 94

View Full Product Details