Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GSA1
Entrez Gene ID
547795
Gene Name
glutamate 1-semialdehyde aminotransferase
Gene Aliases
GSA-AT, Gsa
Chromosome Location
Chr.6: 229292 - 231281 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001251114.1 NP_001238043.1 1 - 2 225 82
GenBank mRNA BT097398.1 - 1 - 2 251 82
L12453.1 - 1 - 2 225 82

Gene Symbol
LOC100790574
Entrez Gene ID
100790574
Gene Name
glutamate-1-semialdehyde 2,1-aminomutase, chloroplastic-like
Gene Aliases
-
Chromosome Location
Chr.4: 189144 - 191067 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK285927.1 - 1 - 2 82

View Full Product Details