Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AAPT1
Entrez Gene ID
732642
Gene Name
aminoalcoholphosphotransferase
Gene Aliases
Cept1
Chromosome Location
Chr.12: 6448848 - 6470795 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001251355.1 NP_001238284.1 11 - 12 868 106
GenBank mRNA AK246054.1 - 11 - 12 823 106
U12735.1 - 11 - 12 868 106

View Full Product Details