Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AAPT1
Entrez Gene ID
732642
Gene Name
aminoalcoholphosphotransferase
Gene Aliases
Cept1
Chromosome Location
Chr.12: 6448848 - 6470795 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001251355.1 NP_001238284.1 17 - 18 1233 70
GenBank mRNA AK246054.1 - 17 - 18 1188 70
U12735.1 - 17 - 18 1233 70

View Full Product Details