Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WRKY23
Entrez Gene ID
100127376
Gene Name
WRKY23 protein
Gene Aliases
-
Chromosome Location
Chr.9: 442507 - 444972 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK245933.1 - 1 - 2 228 80
EU019565.1 - 1 - 2 205 80

View Full Product Details