Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COR1
Entrez Gene ID
100217336
Gene Name
cold-regulated protein
Gene Aliases
-
Chromosome Location
Chr.12: 3433361 - 3436023 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001251697.1 NP_001238626.1 3 - 4 254 69
GenBank mRNA AK245991.1 - 3 - 4 367 69
BT089601.1 - 3 - 4 354 69
FJ393230.1 - 3 - 4 254 69

View Full Product Details