Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GSTa
Entrez Gene ID
547520
Gene Name
2,4-D inducible glutathione S-transferase
Gene Aliases
-
Chromosome Location
Chr.15: 47204506 - 47205868 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001250784.1 NP_001237713.1 2 - 2 647 111
GenBank mRNA AF048978.1 - 2 - 2 647 111

View Full Product Details