Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001226.3 | NP_001217.2 | 5, 6 | 551 | |
NM_032992.2 | NP_116787.1 | 2, 3 | 284 | ||
NR_133012.1 | 4, 5 | 474 | |||
GenBank mRNA | BC000305.2 | - | 5, 6 | 499 | |
BC004460.1 | - | 5, 6 | 499 | 2820468 | |
BM837197.1 | - | 2, 3 | 230 | ||
BX380795.2 | - | 4, 5 | 415 |