Gene Transcripts

Gene Symbol
CHRNG
Entrez Gene ID
1146
Gene Name
cholinergic receptor nicotinic gamma subunit
Gene Aliases
ACHRG
Chromosome Location
Chr. 2: 232539727 - 232546328 on Build GRCh38
UniGene ID
Hs.248101
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005199.4 NP_005190.4 8, 9 938
GenBank mRNA AK125362.1 - 8, 9 938
BC111802.1 - 7, 8 761 40069605

View Full Product Details