Gene Transcripts

Gene Symbol
TIMM8A
Entrez Gene ID
1678
Gene Name
translocase of inner mitochondrial membrane 8 homolog A (yeast)
Gene Aliases
DDP, DDP1, DFN1, MTS, TIM8
Chromosome Location
Chr. X: 101345656 - 101348969 on Build GRCh38
UniGene ID
Hs.447877
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004085.3 NP_004076.1 2 505
GenBank mRNA AK312117.1 - 2 253
BC006994.1 - 2 255 4051368
BC015093.1 - 2 258 3684305
BC070284.1 - 2 238 4281013
CN410182.1 - 2 505
U66035.1 - 2 235

View Full Product Details