Gene Transcripts

Gene Symbol
DUSP7
Entrez Gene ID
1849
Gene Name
dual specificity phosphatase 7
Gene Aliases
MKPX, PYST2
Chromosome Location
Chr. 3: 52048919 - 52056445 on Build GRCh38
UniGene ID
Hs.591664
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001947.3 NP_001938.2 2 884
GenBank mRNA AF508727.1 - 2 652
BC019107.2 - 2 516
BC104880.1 - 2 712 8143883
BC104882.1 - 2 712 8143885
BC128064.1 - 1 2459
X93921.1 - 2 514

View Full Product Details