Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005273.3 | NP_005264.2 | 9 | 1161 | |
GenBank mRNA | AF501883.1 | - | 8 | 888 | |
AK056750.1 | - | 4 | 1283 | ||
AK291592.1 | - | 9 | 1036 | ||
BC010073.2 | - | 9 | 1085 | 4300639 | |
BC012348.1 | - | 9 | 1146 | 4556119 | |
BC068003.1 | - | 9 | 1071 | 6055863 | |
M16538.1 | - | 9 | 1035 | ||
M36429.1 | - | 8 | 945 |