Gene Transcripts

Gene Symbol
GPR4
Entrez Gene ID
2828
Gene Name
G protein-coupled receptor 4
Gene Aliases
Chromosome Location
Chr. 19: 45589764 - 45602208 on Build GRCh38
UniGene ID
Hs.17170
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005282.2 NP_005273.1 2 1933
XM_017026607.1 XP_016882096.1 2 2521
XM_017026608.1 XP_016882097.1 2 1842
GenBank mRNA AK290698.1 - 2 1886
BC038450.1 - 2 1921
BC067535.1 - 1 1292 6971751
BC067536.1 - 1 1292 6971752
EU432116.1 - 1 988

View Full Product Details