Gene Transcripts

Gene Symbol
HIST1H1A
Entrez Gene ID
3024
Gene Name
histone cluster 1, H1a
Gene Aliases
H1.1, H1A, H1F1, HIST1
Chromosome Location
Chr. 6: 26017032 - 26017812 on Build GRCh38
UniGene ID
Hs.150206
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005325.3 NP_005316.1 1 638
GenBank mRNA BC069492.1 - 1 609
BC101593.1 - 1 637 8069099
BC112140.1 - 1 638 8327608

View Full Product Details