Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001258040.2 | NP_001244969.1 | 10 | 1579 | |
NM_001258041.2 | NP_001244970.1 | 11 | 1639 | ||
NM_001258042.2 | NP_001244971.1 | 10 | 1519 | ||
NM_001289092.1 | NP_001276021.1 | 9 | 1477 | ||
NM_001289093.1 | NP_001276022.1 | 8 | 1357 | ||
NM_001289094.1 | NP_001276023.1 | 11 | 1612 | ||
NM_002109.5 | NP_002100.2 | 11 | 1699 | ||
GenBank mRNA | AK000498.1 | - | 11 | 1379 | |
AK094406.1 | - | 2 | 1152 | ||
AK124831.1 | - | 10 | 2889 | ||
AK225776.1 | - | 11 | 1289 | ||
AK293154.1 | - | 9 | 1091 | ||
AK295219.1 | - | 10 | 1175 | ||
AK302295.1 | - | 10 | 1193 | ||
AK303778.1 | - | 8 | 1167 | ||
AK307669.1 | - | 11 | 1292 | ||
AY995220.1 | - | 11 | 1276 | ||
BC011807.2 | - | 11 | 1306 | 4331277 | |
BC080514.1 | - | 11 | 1304 | 2821065 | |
Z11518.1 | - | 11 | 1834 |