Gene Transcripts

Gene Symbol
HK1
Entrez Gene ID
3098
Gene Name
hexokinase 1
Gene Aliases
HK, HK1-ta, HK1-tb, HK1-tc, HKD, HKI, HMSNR, HXK1, hexokinase
Chromosome Location
Chr. 10: 69270000 - 69401882 on Build GRCh38
UniGene ID
Hs.370365
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000188.2 NP_000179.2 9 1149
NM_001322364.1 NP_001309293.1 11 1242
NM_001322365.1 NP_001309294.1 14 1547
NM_001322366.1 NP_001309295.1 9 1040
NM_001322367.1 NP_001309296.1 8 1053
NM_033496.2 NP_277031.1 9 1146
NM_033497.2 NP_277032.1 12 1364
NM_033498.2 NP_277033.1 12 1418
NM_033500.2 NP_277035.2 13 1511
XM_005269737.1 XP_005269794.1 9 1420
XM_011539732.1 XP_011538034.1 13 1418
GenBank mRNA AB209526.1 - 9 1145
AK091267.1 - 9 1274
AK128226.1 - 14 1542
AK292012.1 - 9 1149
AK294428.1 - 9 1146
AK309904.1 - 8 975
BC008730.2 - 9 1104 3163058
M75126.1 - 9 1129
X66957.1 - 3 190

View Full Product Details