Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002254.6 | NP_002245.4 | 7 | 2818 | |
XM_005264299.2 | XP_005264356.2 | 7 | 2815 | ||
GenBank mRNA | AF018164.1 | - | 7 | 2312 | |
AF035621.1 | - | 7 | 2314 | ||
AK293738.1 | - | 8 | 2082 | ||
AK315324.1 | - | 8 | 2842 | ||
BC018932.2 | - | 8 | 794 | ||
BC042486.1 | - | 8 | 2725 | 3866587 | |
BC092406.1 | - | 7 | 2703 | 6504352 | |
BC130423.1 | - | 7 | 2389 | 40146454 | |
BC132783.1 | - | 7 | 2389 | 40146805 | |
BC144223.1 | - | 7 | 2383 | 9052743 | |
BX571741.1 | - | 7 | 2741 |