Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000893.3 | NP_000884.1 | 9 | 1287 | |
NM_001102416.2 | NP_001095886.1 | 9 | 1287 | ||
NM_001166451.1 | NP_001159923.1 | 8 | 1179 | ||
GenBank mRNA | AK223589.1 | - | 9 | 1236 | |
AK290839.1 | - | 9 | 1186 | ||
AK298440.1 | - | 10 | 1201 | ||
AK303768.1 | - | 9 | 1267 | ||
AK315230.1 | - | 9 | 1239 | ||
BC026253.1 | - | 9 | 965 | ||
BC060039.1 | - | 9 | 1190 | 4593077 | |
CB148843.1 | - | 2 | 101 | ||
CD014130.1 | - | 8 | 1141 | ||
K02566.1 | - | 9 | 1102 |