Gene Transcripts

Gene Symbol
LNPEP
Entrez Gene ID
4012
Gene Name
leucyl and cystinyl aminopeptidase
Gene Aliases
CAP, IRAP, P-LAP, PLAP
Chromosome Location
Chr. 5: 96935642 - 97029411 on Build GRCh38
UniGene ID
Hs.527199
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005575.2 NP_005566.2 12 2650
NM_175920.3 NP_787116.2 12 2442
GenBank mRNA AB208883.1 - 11 1524
AK314226.1 - 12 2192
D50810.1 - 12 2607
U62768.1 - 12 2197
U62769.1 - 12 2452

View Full Product Details