Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001204126.1 | NP_001191055.1 | 18 | 2088 | |
NM_001204127.1 | NP_001191056.1 | 18 | 2018 | ||
NM_006152.3 | NP_006143.2 | 19 | 2169 | ||
XM_005253370.3 | XP_005253427.1 | 20 | 2228 | ||
XM_005253372.2 | XP_005253429.1 | 19 | 2229 | ||
XM_005253374.3 | XP_005253431.1 | 17 | 2086 | ||
XM_006719076.2 | XP_006719139.1 | 16 | 1152 | ||
XM_011520667.1 | XP_011518969.1 | 20 | 2377 | ||
XM_011520668.2 | XP_011518970.1 | 16 | 1415 | ||
XM_011520669.2 | XP_011518971.1 | 15 | 1297 | ||
XM_011520670.1 | XP_011518972.1 | 15 | 1068 | ||
XM_017019300.1 | XP_016874789.1 | 19 | 2312 | ||
XM_017019301.1 | XP_016874790.1 | 18 | 2021 | ||
XM_017019302.1 | XP_016874791.1 | 19 | 2226 | ||
GenBank mRNA | AK303103.1 | - | 15 | 1421 | |
AK303256.1 | - | 18 | 1627 | ||
BC126417.1 | - | 19 | 1676 | 8992133 | |
BC144164.1 | - | 18 | 1595 | ||
U10485.1 | - | 19 | 1817 |