Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005372.1 | NP_005363.1 | 1 | 250 | |
GenBank mRNA | BC069569.1 | - | 1 | 250 | |
BC069590.1 | - | 1 | 250 | ||
BC106737.1 | - | 1 | 250 | 40016104 | |
BC106738.1 | - | 1 | 250 | 40016106 |