Gene Transcripts

Gene Symbol
SLC11A2
Entrez Gene ID
4891
Gene Name
solute carrier family 11 member 2
Gene Aliases
AHMIO1, DCT1, DMT1, NRAMP2
Chromosome Location
Chr. 12: 50952263 - 51028335 on Build GRCh38
UniGene ID
Hs.505545
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000617.2 NP_000608.1 13 1385
NM_001174125.1 NP_001167596.1 13 1377
NM_001174126.1 NP_001167597.1 13 1385
NM_001174127.1 NP_001167598.1 13 1380
NM_001174128.1 NP_001167599.1 13 1380
NM_001174129.1 NP_001167600.1 13 1645
NM_001174130.1 NP_001167601.1 12 1360
NR_033421.1 13 1305
NR_033422.1 13 1380
XM_005268911.3 XP_005268968.1 13 1437
XM_005268912.4 XP_005268969.1 12 1562
XM_011538404.2 XP_011536706.1 13 1761
XM_011538405.2 XP_011536707.1 13 1369
XM_017019355.1 XP_016874844.1 13 1645
XM_017019356.1 XP_016874845.1 13 1575
GenBank mRNA AB004857.1 - 13 1328
AB062284.1 - 13 1302
AF064484.1 - 13 1328
AJ493662.1 - 13 1377
AK094735.1 - 13 1645
AK128640.1 - 13 1277
AK296445.1 - 12 1339
AK315987.1 - 13 1291
AK316507.1 - 12 1316
AL832886.1 - 3 711
BC002592.2 - 13 1332 3160522
BC100014.1 - 13 1353 5300266
KT455015.1 - 13 1341

View Full Product Details