Gene Transcripts

Gene Symbol
PAH
Entrez Gene ID
5053
Gene Name
phenylalanine hydroxylase
Gene Aliases
PH, PKU, PKU1
Chromosome Location
Chr. 12: 102838326 - 102917603 on Build GRCh38
UniGene ID
Hs.603740
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000277.1 NP_000268.1 7 1222
GenBank mRNA AK313383.1 - 8 847
BC026251.1 - 7 903 4767736
K03020.1 - 7 972
L47726.1 - 7 1222
U49897.1 - 7 1222

View Full Product Details