Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002740.5 | NP_002731.4 | 12 | 1359 | |
XM_017006860.1 | XP_016862349.1 | 11 | 1266 | ||
GenBank mRNA | AB451443.1 | - | 12 | 1094 | |
AK315342.1 | - | 12 | 1287 | ||
BC022016.2 | - | 12 | 1354 | 4823853 | |
L18964.1 | - | 12 | 1358 | ||
L33881.1 | - | 12 | 1298 |