Gene Transcripts

Gene Symbol
PTX3
Entrez Gene ID
5806
Gene Name
pentraxin 3
Gene Aliases
TNFAIP5, TSG-14
Chromosome Location
Chr. 3: 157436791 - 157443628 on Build GRCh38
UniGene ID
Hs.591286
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002852.3 NP_002843.2 3 990
GenBank mRNA AK312705.1 - 3 990
BC039733.1 - 3 963 5768379
M31166.1 - 3 920
X63053.1 - 3 852
X63613.1 - 3 914

View Full Product Details