Gene Transcripts

Gene Symbol
PEX2
Entrez Gene ID
5828
Gene Name
peroxisomal biogenesis factor 2
Gene Aliases
PAF1, PBD5A, PBD5B, PMP3, PMP35, PXMP3, RNF72, ZWS3
Chromosome Location
Chr. 8: 76980258 - 77001044 on Build GRCh38
UniGene ID
Hs.437966
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000318.2 NP_000309.1 4 631
NM_001079867.1 NP_001073336.1 3 599
NM_001172086.1 NP_001165557.1 5 637
NM_001172087.1 NP_001165558.1 3 495
GenBank mRNA AK312997.1 - 2 298
BC000661.2 - 4 404 3347824
BC005375.1 - 3 400 3934225
BC093043.1 - 4 417 30399827
BP193912.1 - 3 398
M85038.1 - 4 438
M86852.1 - 4 546

View Full Product Details