Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001136026.2 | NP_001129498.1 | 9 | 1053 | |
NM_001136232.2 | NP_001129704.1 | 8 | 907 | ||
NM_030673.3 | NP_109598.2 | 9 | 909 | ||
NM_183352.2 | NP_899195.1 | 8 | 875 | ||
XM_005265379.2 | XP_005265436.1 | 9 | 926 | ||
XM_017007019.1 | XP_016862508.1 | 9 | 963 | ||
XM_017007020.1 | XP_016862509.1 | 8 | 770 | ||
XM_017007021.1 | XP_016862510.1 | 8 | 780 | ||
GenBank mRNA | AB209554.1 | - | 7 | 722 | |
AF052155.1 | - | 8 | 907 | ||
AK095629.1 | - | 6 | 2734 | ||
AK223019.1 | - | 8 | 760 | ||
AK301999.1 | - | 9 | 977 | ||
BC002634.2 | - | 9 | 790 | 3605545 | |
BC006167.1 | - | 5 | 567 | ||
BC091506.1 | - | 8 | 785 | 30408210 | |
L09260.1 | - | 8 | 752 |