Gene Transcripts
- Gene Symbol
- SLC11A1
- Entrez Gene ID
- 6556
- Gene Name
- solute carrier family 11 member 1
- Gene Aliases
- LSH, NRAMP, NRAMP1
- Chromosome Location
- Chr. 2: 218381766 - 218396894 on Build GRCh38
- UniGene ID
- Hs.591607
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000578.3 |
NP_000569.3
|
9, 10 |
1290 |
|
XM_005246793.3 |
XP_005246850.1
|
8, 9 |
1410 |
|
XM_005246794.3 |
XP_005246851.1
|
10, 11 |
1627 |
|
XM_006712709.3 |
XP_006712772.1
|
9, 10 |
1484 |
|
XM_006712710.3 |
XP_006712773.1
|
8, 9 |
1361 |
|
XM_006712711.3 |
XP_006712774.1
|
7, 8 |
1287 |
|
XM_011511684.2 |
XP_011509986.1
|
10, 11 |
1662 |
|
XM_011511685.2 |
XP_011509987.1
|
9, 10 |
1519 |
|
XM_017004765.1 |
XP_016860254.1
|
8, 9 |
1430 |
|
XM_017004766.1 |
XP_016860255.1
|
9, 10 |
1521 |
|
GenBank mRNA |
AB209650.1 |
- |
7, 8 |
3004 |
|
AK298667.1 |
- |
9, 10 |
1044 |
|
AK303398.1 |
- |
9, 10 |
1014 |
|
AK307954.1 |
- |
9, 10 |
1230 |
|
AK310914.1 |
- |
9, 10 |
1046 |
|
AK311405.1 |
- |
8, 9 |
897 |
|
BC033754.1 |
- |
7, 8 |
1296 |
|
BC071165.1 |
- |
10, 11 |
1084 |
5756382 |
D38171.1 |
- |
7, 8 |
797 |
|
D50402.1 |
- |
9, 10 |
1125 |
|
D50403.1 |
- |
9, 10 |
1036 |
|
L32185.1 |
- |
9, 10 |
1026 |
|
View Full Product Details