Gene Transcripts
- Gene Symbol
- CAPN15
- Entrez Gene ID
- 6650
- Gene Name
- calpain 15
- Gene Aliases
- SOLH
- Chromosome Location
- Chr. 16: 527795 - 554636 on Build GRCh38
- UniGene ID
- Hs.632219
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_005632.2 |
NP_005623.1
|
7, 8 |
2418 |
|
XM_011522620.2 |
XP_011520922.1
|
8, 9 |
5717 |
|
XM_011522621.2 |
XP_011520923.1
|
8, 9 |
6645 |
|
XM_011522622.1 |
XP_011520924.1
|
6, 7 |
5349 |
|
XM_011522623.2 |
XP_011520925.1
|
9, 10 |
3420 |
|
XM_011522624.2 |
XP_011520926.1
|
8, 9 |
2758 |
|
XM_011522625.2 |
XP_011520927.1
|
7, 8 |
2644 |
|
XM_011522626.2 |
XP_011520928.1
|
8, 9 |
6209 |
|
XM_011522627.2 |
XP_011520929.1
|
8, 9 |
6214 |
|
XM_011522628.2 |
XP_011520930.1
|
8, 9 |
3278 |
|
XM_011522629.2 |
XP_011520931.1
|
8, 9 |
3141 |
|
XM_011522630.2 |
XP_011520932.1
|
7, 8 |
2479 |
|
XM_011522631.2 |
XP_011520933.1
|
6, 7 |
2365 |
|
XM_011522632.2 |
XP_011520934.1
|
6, 7 |
2796 |
|
XM_017023596.1 |
XP_016879085.1
|
9, 10 |
3557 |
|
GenBank mRNA |
BC021854.1 |
- |
3, 4 |
485 |
|
U85647.1 |
- |
7, 8 |
2418 |
|
View Full Product Details