Gene Transcripts
- Gene Symbol
- SUPT4H1
- Entrez Gene ID
- 6827
- Gene Name
- SPT4 homolog, DSIF elongation factor subunit
- Gene Aliases
- SPT4, SPT4H, SUPT4H, Supt4a
- Chromosome Location
- Chr. 17: 58345175 - 58352238 on Build GRCh38
- UniGene ID
- Hs.439481
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_003168.2 |
NP_003159.1
|
3, 4 |
328 |
|
NR_073470.1 |
|
3, 4 |
330 |
|
GenBank mRNA |
AK129758.1 |
- |
3, 4 |
299 |
|
AK311986.1 |
- |
3, 4 |
241 |
|
BC002802.2 |
- |
3, 4 |
286 |
3636217 |
BP196741.1 |
- |
3, 4 |
328 |
|
BP318625.1 |
- |
3, 4 |
291 |
|
BX331655.2 |
- |
3, 4 |
292 |
|
CR407663.1 |
- |
3, 4 |
225 |
|
U38817.1 |
- |
3, 4 |
267 |
|
U38818.1 |
- |
3, 4 |
286 |
|
U43923.1 |
- |
3, 4 |
292 |
|
View Full Product Details