Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003214.3 | NP_003205.2 | 4 | 521 | |
GenBank mRNA | AF142482.1 | - | 4 | 475 | |
AK226144.1 | - | 1 | 886 | ||
BC010053.2 | - | 4 | 377 | ||
BC091488.1 | - | 4 | 446 | 6187474 | |
BQ719548.1 | - | 4 | 445 | ||
DA856821.1 | - | 4 | 544 | ||
X94439.1 | - | 3 | 285 |