Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001199849.1 | NP_001186778.1 | 10 | 968 | |
NM_001199850.1 | NP_001186779.1 | 8 | 773 | ||
NM_001199851.1 | NP_001186780.1 | 8 | 752 | ||
NM_004632.3 | NP_004623.1 | 9 | 872 | ||
NM_033657.2 | NP_387506.1 | 9 | 941 | ||
XM_005245480.2 | XP_005245537.1 | 8 | 838 | ||
XM_005245481.2 | XP_005245538.1 | 8 | 817 | ||
XM_005245482.4 | XP_005245539.1 | 7 | 652 | ||
XM_011509972.2 | XP_011508274.1 | 8 | 790 | ||
XM_017002289.1 | XP_016857778.1 | 8 | 859 | ||
XM_017002290.1 | XP_016857779.1 | 9 | 883 | ||
XM_017002291.1 | XP_016857780.1 | 9 | 858 | ||
XM_017002292.1 | XP_016857781.1 | 9 | 837 | ||
XM_017002293.1 | XP_016857782.1 | 8 | 756 | ||
XM_017002294.1 | XP_016857783.1 | 8 | 777 | ||
XM_017002295.1 | XP_016857784.1 | 8 | 756 | ||
XM_017002296.1 | XP_016857785.1 | 7 | 736 | ||
XM_017002297.1 | XP_016857786.1 | 7 | 715 | ||
GenBank mRNA | AK225869.1 | - | 10 | 967 | |
AK293117.1 | - | 9 | 873 | ||
AK298201.1 | - | 8 | 760 | ||
AK302281.1 | - | 8 | 773 | ||
BC002358.1 | - | 8 | 900 | ||
BC107487.1 | - | 9 | 872 | 40032516 | |
BC107488.1 | - | 9 | 941 | 40032512 | |
BT019494.1 | - | 8 | 784 | ||
CR749790.1 | - | 9 | 952 | ||
U18321.1 | - | 9 | 953 | ||
X83544.1 | - | 9 | 857 |