Gene Transcripts

Gene Symbol
OR1A1
Entrez Gene ID
8383
Gene Name
olfactory receptor family 1 subfamily A member 1
Gene Aliases
OR17-7
Chromosome Location
Chr. 17: 3215621 - 3216550 on Build GRCh38
UniGene ID
Hs.532688
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014565.2 NP_055380.2 1 771
GenBank mRNA BC069169.1 - 1 771
BC128248.1 - 1 771
BC141934.1 - 1 771 40114773
BC141935.1 - 1 771 40114774

View Full Product Details